Supelco® TraceCERT® Organic Reference Standard, BTEX Mix, Underground Storage Tank
Catalog # CRM48026
Supplier: Merck
CAS Number:
Specifications
- Concentration:200 μg/ml each component in methanol
- Pk:1 mL
- Standards:Certified reference material, TraceCERT®
- Description:BTEX mix, underground storage tank
- Storage conditions:2…8 °C
- Hazard class description:H225 - H301 + H311 + H331 - H370
- Shipping conditions:WICE
- M-Clarity quality level:MQ100
- Synonym(s):BTEX Solution; UST BTEX Mix
- Flash point:9,7 °C - Closed cup
Specifications
About this item
- GHS signal: GHS02, GHS06, GHS08
Caution: P210 - P233 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311