
Sodium hydroxide 5.0 M NaOH (5.0N) volumetric solution, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Boiling Pt:105…140 °C (1.013 hPa)
Melting Pt:–12…–10 °C
Density:1.327 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:1824
ADR:8,II
:GHS05
Specification Test Results
- Assay:4.900 - 5.100 N
- Traceable to NIST SRM:effective
- Vapor Pressure:3 mmHg ( 37 °C)
Specification Test Results
Frequently Bought Together