Calbiochem® Bovine native Fibrinogen (from Plasma)
Catalog # 341573-1
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Förpackningstyp:Glass bottle
- Förp.:1 g
- CAS-nr:9001-32-5
- Protein/peptide type:Native
- Källa:Plasma
- Species:Bovine
- Storage conditions:2...8 °C
- Form:Lyophilised
- Protein/peptide name:Fibrinogen
- Formulering:Lyophilised from 20 mM sodium citrate-HCl, pH 7,4
Specifications
About this item
Fibrinogen is a soluble plasma glycoprotein that is synthesised in the hepatic parenchymal cells.
- Contains more than 95% clottable protein