Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Poloxamer 407 (Pluronic® F-127)
Pluronic® F-127 20% in DMSO
Pluronic® F-127 20% in DMSO
Pluronic® F-127 20% in DMSO
Catalog # 59004.
Supplier:
Biotium
CAS Number:
Expand image
Pluronic® F-127 20% in DMSO
Catalog # 59004.
Supplier:
Biotium
CAS Number:
Specifications
Pk:
1 mL
Seq:
2070
Specifications
About this item
Product Details
:
CAS Number:
9003-11-6
Formula:
HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)aH
MDL Number:
MFCD00082049
Product Details