Order Entry
Sweden
ContactUsLinkComponent
Supelco® TraceCERT® Organic Reference Standard, EPA 8270 Ether/Phthalate Mix
Supelco® TraceCERT® Organic Reference Standard, EPA 8270 Ether/Phthalate Mix
Catalog # CRM47643
Supplier:  Merck
CAS Number:  
Supelco® TraceCERT® Organic Reference Standard, EPA 8270 Ether/Phthalate Mix
Catalog # CRM47643
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Certified reference material, TraceCERT®
  • Description:
    EPA 8270 Ether/Phthalate mix
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H315 - H319 - H336 - H351 - H412
  • Shipping conditions:
    WICE
  • M-Clarity quality level:
    MQ100

Specifications

About this item

  • GHS signal: GHS08, GHS07, GHS09
Caution: P202 - P261 - P273 - P302 + P352 - P305 + P351 + P338 - P308 + P313