Order Entry
Sweden
ContactUsLinkComponent
Supelco® Organic Reference Standard, Aldrin
Supelco® Organic Reference Standard, Aldrin
Katalog # 08573-50MG
Leverantör:  Merck
CAS-nummer:  
undefined
Supelco® Organic Reference Standard, Aldrin
Katalog # 08573-50MG
Leverantör:  Merck
CAS-nummer:  

Specifikationer

  • Pk:
    50 mg
  • Standards:
    Certified reference material, TraceCERT®
  • CAS No.:
    309-00-2
  • Description:
    Aldrin
  • Storage conditions:
    –20 °C
  • Hazard class description:
    H300 + H310 - H351 - H372 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00135587
  • M-Clarity quality level:
    MQ300
  • Molecular weight:
    364,91
  • Flash point:
    Not applicable

Specifikationer

Om denna produkt

  • GHS signal: GHS06, GHS08, GHS09
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Caution: P202 - P264 - P273 - P280 - P301 + P310 - P302 + P352 + P310