Order Entry
Sweden
ContactUsLinkComponent
Coomassie Brilliant Blue Destaining Solution, G-Biosciences
Coomassie Brilliant Blue Destaining Solution, G-Biosciences
Catalog # 786-499
Supplier:  G-Biosciences
CAS Number:  
undefined
Coomassie Brilliant Blue Destaining Solution, G-Biosciences
Catalog # 786-499
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pk:
    1 L
  • Description:
    Coomassie Brilliant Blue Destaining Solution
  • Seq:
    03

Specifications

About this item

After Coomassie Brilliant Blue staining process, the band intensity may be further enhanced by de-staining the stained gel in our CBB De-Staining Solution.

  • Contains Methanol and Acetic acid