Coomassie® brilliant blue R-250 solution electrophoresis gel stain, ready-to-use
Catalog # 786-498
Supplier: G-Biosciences
CAS Number:
Specifications
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:825.98 g/mol
Storage Temperature:Ambient
EINECS:228-058-4
CAS Number:6104-59-2
Formula:C₄₅H₄₄N₃NaO₇S₂
MDL Number:MFCD00041762