Order Entry
Sweden
ContactUsLinkComponent
6-Carboxy-X-rhodamine triethylammonium salt single isomer ≥95% (by TLC) amine-reactive fluorescent dye
6-Carboxy-X-rhodamine triethylammonium salt single isomer ≥95% (by TLC) amine-reactive fluorescent dye
Catalog # 90007.
Supplier:  Biotium
CAS Number:  
undefined
6-Carboxy-X-rhodamine triethylammonium salt single isomer ≥95% (by TLC) amine-reactive fluorescent dye
Catalog # 90007.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :635.6 g/mol
Storage Temperature:Refrigerator
Formula:C₃₉H₄₅N₃O₅