Food standards, Fusarenon X, neat, Supelco®
33438-5MG
: Merck
:
- Koncentration:Neat
- Förp.:5 mg
- CAS-nr:23255-69-8
- Beskrivning:Fusarenon X
- M-Clarity quality level:MQ100
- Benämning:Fusarenon X
Fusarenon X is a trichothecene mycotoxin isolated from Fusarium nivale.
Fusarenon X may be used as a reference standard for the determination of fusarenon X in barley, wheat and corn samples by high-performance thin-layer chromatography (HPTLC).
: Storage temperature: Freezer −20 °C