NMR tubes, polyethylene pressure caps
Leverantör: DWK Life Sciences
Certifikat
Om denna produkt
Tight-fitting, polyethylene cap which stays in place even with a build-up of internal vaporisation pressures.
Tight-fitting, polyethylene cap which stays in place even with a build-up of internal vaporisation pressures.