Industrial gloves, uvex compact NB27E
: UVEX
These robust cotton jersey gloves feature a special NBR coating on palm, fingers and ³/₄ of the back of the hand. They are ideal for rough work and handling coarse materials.
- Very good mechanical abrasion resistance
- High tear resistance
Areas of application: Iron and steel industry, shipping, logistics, concrete and construction work, wood processing.
: EN 388:2016 (4121X)
- Catalog No:
- UVEX6094610
- UVEX6094609
- Model:
- NB27E
- NB27E
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Liner material:
- Cotton jersey
- Cotton jersey
- Finish:
- Palm and fingers coated
- Palm and fingers coated
- Cuff style:
- Gauntlet
- Gauntlet
- EN norm:
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)