Welding spectacles, 9104
Catalog # UVEX9104.041
Supplier: UVEX
Specifications
- Type:Anti-UV, scratch-resistant, welding level 1.7
- Colour:Black/green
- Seq:01
- Lenses:Clear/grey PC
- Pk:1
Specifications
About this item
Compact flip-up welding safety spectacles with a close fit and anatomically shaped nose bridge, side shields and integrated brow guard are designed for increased protection with flip-up and easily replaceable lens.
- Perfect colour recognition thanks to uvex sunglare filters
- Reliable UV 400 and IR protection (flip-up lens)
- Minimum damage from welding sparks
- Side protection and integrated brow guard frame design
- Fully adjustable side arms
- Adjustable, soft and comfortable ear pieces
The specially moulded frame edge provides protection against particles from below
Certifications: EN 166, EN 169, EN 170