Order Entry
Sweden
ContactUsLinkComponent
Hexamethylenetetramine, white crystalline powder ACS
Hexamethylenetetramine, white crystalline powder ACS
Supplier:  MP Biomedicals
Certificates
undefined
Hexamethylenetetramine, white crystalline powder ACS
Supplier:  MP Biomedicals

Specification Test Results

  • Assay (dried basis):
    >99%
  • Loss on Drying:
    <2.0%
  • Residue after Ignition:
    <0.1%
  • Heavy Metals (as Pb):
    <0.001%

Specification Test Results