Cut resistant gloves, uvex synexo impact 1
Supplier: UVEX
Certificates
About this item
These HPPE/glass fibre cut protection gloves with NBR coating and Grip finish on palm and fingertips combine the benefits of seamless safety gloves with specially designed protection zones.
- Protection against shocks and impacts thanks to the extra padding in the palm area
- Very high level of cut protection
- Good grip in dry and damp areas
- Good fit, high flexibility, good wearer comfort
- Flexible velcro fastening
- Protectors on the back of the hand and reinforcements on the finger joints
Ideal for use in tasks with extreme mechanical stress, handling heavy-duty tools, building and consctruction industry, mining, drilling, oil and gas industry.
Certifications: EN 388:2016 (4X43CP)
Specifications
- Catalog No:
- UVEX6059810
- UVEX6059811
- Model:
- impact 1
- impact 1
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Liner material:
- HPPE/glass fibre
- HPPE/glass fibre
- Finish:
- NBR coating and Grip finish on palm and fingertips
- NBR coating and Grip finish on palm and fingertips
- Cuff style:
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4X43CP)
- EN 388:2016 (4X43CP)