Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Acesulphame potassium
Acesulphame potassium ≥99%
Acesulphame potassium ≥99%
Acesulphame potassium ≥99%
Leverantör:
Spectrum Chemical
Certifikat
Expand image
Acesulphame potassium ≥99%
Leverantör:
Spectrum Chemical
Om denna produkt
CAS-nummer:
55589-62-3
:
Formel:
C₄H₄KNO₄S
MDL-nummer:
MFCD00043833
:
201.24 g/mol
Specifikationstestresultat
Assay:
99%
Specifikationstestresultat