Order Entry
Sweden
ContactUsLinkComponent
Calcofluor white stain fluorescent dye, for microbiology, Sigma-Aldrich®
Irritant
Health
Calcofluor white stain fluorescent dye, for microbiology, Sigma-Aldrich®
Catalog # 18909-100ML-F
CAS Number:  
Irritant
Health
Calcofluor white stain fluorescent dye, for microbiology, Sigma-Aldrich®
Catalog # 18909-100ML-F
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :916.98 g/mol
 :M-Clarity™ quality level = MQ100
 :GHS07
 :GHS08
Formula:C₄₀H₄₄N₁₂O₁₀S₂