Order Entry
Sweden
ContactUsLinkComponent
Azo rubine (50% dye content) C.I 14720 , Sigma-Aldrich®
Azo rubine (50% dye content) C.I 14720 , Sigma-Aldrich®
Catalog # 214515-50G
CAS Number:  
undefined
Azo rubine (50% dye content) C.I 14720 , Sigma-Aldrich®
Catalog # 214515-50G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 g
  • Seq:
    3368

Specifications

Specification Test Results

  • Infrared Spectrum:
    Conforms to Structure
  • Appearance (Color), Dark Red to Brown to Black:
    Conforms to Requirements
  • Wavelength:
    513 - 519 nm
  • Extinction Coefficient:
    ≥14500
  • Extinction Coefficient, c = 0.03 g/l, H₂O:
    ≥33000
  • Carbon:
    23.9 - 71.7%
  • Nitrogen:
    2.7 - 8.4%

Specification Test Results

About this item

 :
 :502.42 g/mol
 :M-Clarity™ quality level = MQ200
Flash Pt:>225 °C
EINECS:222-657-4
CAS Number:3567-69-9
Formula:C₂₀H₁₄N₂O₇S₂
MDL Number:MFCD00003978