Diaion® SA10A (Cl) 1,3 meq/ml på poly (styren-divinylbensen), cross-linked, beads 300 - 1180 µm, strongly basic gel type ion-exchange resin
Katalog # 46796.A1
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Förp.:1 kg
- Seq:4024
Specifications
About this item
:
Storage Temperature:Omgivande