Order Entry
Sweden
ContactUsLinkComponent
Bovine mucin (from Submaxillary glands)
Bovine mucin (from Submaxillary glands)
Catalog # ABNOP5239
Supplier:  Abnova
CAS Number:  
undefined
Bovine mucin (from Submaxillary glands)
Catalog # ABNOP5239
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Source:
    Submaxillary glands
  • Species:
    Bovine
  • Storage conditions:
    4 °C
  • Form:
    Lyophilised
  • Protein/peptide name:
    Mucin

Specifications

About this item

Native protein purified from bovine submaxillary gland used as a enzyme substrate.

Mucins are the glycoproteins in epithelial mucous secretions, their high viscosity provide protective function to the epithelial cell. Suitable as a substrate for neuraminidase.