Order Entry
Sweden
ContactUsLinkComponent
Zinc chloride ≥98%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
Zinc chloride ≥98%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
Leverantör:  Honeywell Chemicals
Certifikat
Zinc chloride ≥98%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
Leverantör:  Honeywell Chemicals
Certifikat

Specifikationstestresultat

  • Assay:
    98.0 - 100.5 %
  • Insoluble in HCl:
    Max. 0.005 %
  • pH (10 %, 20°C):
    4.6 - 5.5
  • Ammonium (NH₄):
    Max. 50 ppm
  • Calcium (Ca):
    Max. 0.001 %
  • Cadmium (Cd):
    Max. 0.0005 %
  • Copper (Cu):
    Max. 0.001 %
  • Iron (Fe):
    Max. 0.0005 %
  • Potassium (K):
    Max. 0.001 %
  • Magnesium (Mg):
    Max. 0.001 %
  • Sodium (Na):
    Max. 0.001 %
  • Lead (Pb):
    Max. 0.001 %
  • Al, Ca, Fe, Mg and heavy metals:
    complying
  • Oxide chloride (as ZnO):
    Max. 1.2 %
  • Nitrate (NO₃):
    Max. 0.003 %
  • Sulfate (SO₄):
    Max. 20 ppm
  • Total N:
    Max. 0.002 %

Specifikationstestresultat

Relaterad information