mjölksyra ≥99%, tested according to Ph. Eur., Fluka™
Leverantör: Honeywell Chemicals
Certifikat
Om denna produkt
CAS-nummer:50-21-5
:598-82-3
Formel:H₃CCH(OH)COOH
MDL-nummer:MFCD00004520
Merck Index:13,05351
Kokande Pt:122 °C (20 hPa)
Densitet:1,209 g/cm³
Flash Pt:113 °C
:90.08 g/mol
:GHS05
Förvaringstemperatur:Omgivande
Specifikationstestresultat
- Appearance of the substance:complying
- Assay of total acid:88.0 - 92.0 %
- Sulfated ash:Max. 0.1 %
- Calcium (Ca):Max. 200 ppm
- Heavy metals (as Pb):Max. 10 ppm
- Sulfate (SO₄):Max. 200 ppm
- Citronellic-, Oxalic- and Phosphoric acid:complying
- Sugar and other reducing substances:complying
- In ether insoluble matter:complying
- Residual solvents:complying