Eriochrome Black T Reag. Ph. Eur. indicator for metal titration, Fluka™
Katalog # 32751-100G
Leverantör: Honeywell Chemicals
CAS-nummer:
Specifikationer
- Förpackningstyp:Glass bottle
- Förp.:100 g
- Seq:3401
Specifikationer
Specifikationstestresultat
- Identity (IR):complying
- Solubility in methanol:complying
- Indicator solution:complying
- Suitability for Complexometry:complying
- UV spectrum:complying
Specifikationstestresultat
Om denna produkt
:
Merck Index:13,03702
Förvaringstemperatur:Omgivande
ADR:9,III
:461.39 g/mol
EINECS:217-250-3
CAS-nummer:1787-61-7
Formel:C₂₀H₁₂N₃NaO₇S
MDL-nummer:MFCD00003935