0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Iron (III) oxide
Iron(III) oxide ≥95%, purified, Fluka™
Iron(III) oxide ≥95%, purified, Fluka™
:
Honeywell Chemicals
Iron(III) oxide ≥95%, purified, Fluka™
:
Honeywell Chemicals
:
1309-37-1
:
:
Fe₂O₃
:
MFCD00011008
:
159.69 g/mol
:
13,04058
:
1.565 °C
:
5 g/cm³ (20 °C)
:
Ambient
:
215-168-2
Assay:
Min. 95 %
Loss on ignition:
Max. 0.6 %
sds_12342