
Ammonium sulphamate ≥98.0% ACS, Fluka™
228745-1KG
:
- Pack type:Plastic bottle
- Pk:1 kg
- Seq:4024
- Identity (IR):complying
- Assay:Min. 98.0 %
- Melting point:131.0 - 135.0 °C
- Residue on ignition:Max. 0.1 %
- Water insoluble matter:Max. 0.02 %
- Heavy metals (as Pb):Max. 5 ppm
:
:114.13 g/mol
:12,00589
:131-135 °C
:1.00 g/cm³
:231-871-7
:GHS07
:Ambient
:7773-06-0
:H₆N₂O₃S
:MFCD00011429
:131…135 °C