Order Entry
Sweden
ContactUsLinkComponent
Potassium hydroxide ≥85%, pellets, Puriss. p.a. Reag. Ph. Eur., Fluka™
Potassium hydroxide ≥85%, pellets, Puriss. p.a. Reag. Ph. Eur., Fluka™
Certificates
Potassium hydroxide ≥85%, pellets, Puriss. p.a. Reag. Ph. Eur., Fluka™
Certificates

Specification Test Results

  • Assay:
    85.0 - 100.5 %
  • Assay of K₂CO₃:
    Max. 1.0 %
  • Aluminium (Al):
    Max. 10 ppm
  • Calcium (Ca):
    Max. 0.001 %
  • Iron (Fe):
    Max. 5 ppm
  • Sodium (Na):
    Max. 0.5 %
  • Nickel (Ni):
    Max. 0.0005 %
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Chloride (Cl):
    Max. 100 ppm
  • Phosphate (PO₄):
    Max. 5 ppm
  • Silicate (as SiO₂):
    Max. 0.005 %
  • Sulfate (SO₄):
    Max. 100 ppm
  • Total N:
    Max. 0.0003 %

Specification Test Results

Related Information