Order Entry
Sweden
ContactUsLinkComponent
tert-Butyl methyl ether ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Flammable
Irritant
tert-Butyl methyl ether ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Catalog # 650560-1L
CAS Number:  
undefined
Flammable
Irritant
tert-Butyl methyl ether ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Catalog # 650560-1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Purity (GC):
    Min. 99.85 %
  • IR spectrum:
    complying
  • Non-volatile matter:
    Max. 0.0001 %
  • Water (Karl Fischer):
    Max. 0.02 %
  • Absorbance at 400 nm:
    Max. 0.005
  • Absorbance at 300 nm:
    Max. 0.005
  • Absorbance at 250 nm:
    Max. 0.10
  • Absorbance at 225 nm:
    Max. 0.50
  • Absorbance at 210 nm:
    Max. 1.0
  • Fluorescence (chinin) at 365 nm:
    Max. 1.0 ppb
  • GC (as heptachlor epoxide):
    Max. 10 ng/l
  • Refractive Index:
    n20/D 1.369 (lit.)
  • Autoignition Temperature:
    705 °F

Specification Test Results

About this item

 :
 :88.15 g/mol
Merck Index:13,06059
Boiling Pt:55.3 °C (1.013 hPa)
Melting Pt:-109 °C
Melting Pt:–109 °C
Density:0.74 g/cm³ (20 °C)
Flash Pt:8.5 %(V)
Storage Temperature:Ambient
EINECS:216-653-1
UN:2398
ADR:3,II
 :GHS02
 :GHS07
CAS Number:1634-04-4
Formula:(CH₃)₃COCH₃
MDL Number:MFCD00008812