Order Entry
Sweden
ContactUsLinkComponent
 
Sodium nitrite ≥99%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
 
Sodium nitrite ≥99%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
 

 

  • Assay:
    Min. 99.0 %
  • Water insoluble matter:
    Max. 0.005 %
  • Calcium (Ca):
    Max. 0.002 %
  • Iron (Fe):
    Max. 0.001 %
  • Potassium (K):
    Max. 0.005 %
  • Heavy metals (as Pb):
    Max. 0.001 %
  • Chloride (Cl):
    Max. 0.005 %
  • Sulfate (SO₄):
    Max. 0.005 %