Sodium periodate ≥99.8%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Leverantör: Honeywell Chemicals
Certifikat
Om denna produkt
CAS-nummer:7790-28-5
:
Formel:NaIO₄
MDL-nummer:MFCD00003534
Merck Index:13,08714
Smält Pt:270 °C (dec.)
Densitet:3.86 g/cm³
Förvaringstemperatur:Ambient
EINECS:232-197-6
FN:3085
ADR:5.1,I
:213.89 g/mol
:GHS05
:GHS08
:GHS03
Specifikationstestresultat
- Assay:Min. 99.8 %
- Assay (excluding dried substances):99.8 - 100.3 %
- Loss on drying (110°C):Max. 0.2 %
- Iron (Fe):Max. 0.001 %
- Manganese (Mn):Max. 0.0001 %
- Heavy metals (as Pb):Max. 0.001 %
- Other halogens (as Cl):Max. 0.01 %
- Sulfate (SO₄):Max. 0.005 %