Acetic acid ≥99.7% ACS, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:64-19-7
:
Formula:H₃CCOOH
MDL Number:MFCD00036152
:60.05 g/mol
Boiling Pt:118 °C (1.013 hPa)
Melting Pt:16 °C
Density:1.05 g/cm³ (20 °C)
Flash Pt:40 °C
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
ADR:8,II
REACH:01-2119475328-30
:GHS02
:GHS05
Specification Test Results
- Appearance:complying
- Identity (IR):complying
- Assay (GC):Min. 99.7 %
- Non-volatile matter:Max. 0.001 %
- Titrable Base:Max. 0.0004 meq/g
- Iron (Fe):Max. 0.00002 %
- Heavy metals (as Pb):Max. 0.00005 %
- Chloride (Cl):Max. 0.0001 %
- Sulfate (SO₄):Max. 0.0001 %
- Acetic aNHydride (GC):Max. 0.01 %
- KMnO₄ reducing substances:complying
- K₂Cr₂O₇ reducing matter:complying
- APHA:Max. 10
- Mixable with H₂O:complying
- Explosion Limit:16 %, 92 °F
- Refractive Index:n20/D 1.371 (lit.)