Order Entry
Sweden
ContactUsLinkComponent
Acetic acid solution 49-51% (by titrimetric analysis) in water for HPLC, Fluka™
Acetic acid solution 49-51% (by titrimetric analysis) in water for HPLC, Fluka™
Leverantör:  Honeywell Chemicals
Certifikat
undefined
Acetic acid solution 49-51% (by titrimetric analysis) in water for HPLC, Fluka™
Leverantör:  Honeywell Chemicals

Specifikationstestresultat

  • assay (HPLC):
    Min. 99 %
  • water (Karl Fischer):
    Max. 0.5 %
  • identity (NMR):
    complying
  • melting range:
    171 - 175 °C
  • Absorbance at 300 nm:
    Max. 0.01
  • Absorbance at 270 nm:
    Max. 0.02
  • Absorbance at 260 nm:
    Max. 0.05

Specifikationstestresultat