Analytical standard, 4′-Hydroxydiclofenac, reference material, Honeywell Fluka™
HONCR1336-10MG
:
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:64118-84-9
- Description:4′-Hydroxydiclofenac
- Melting point:175…176 °C
- Formula:C₁₄H₁₁Cl₂NO₃
- Storage temperature:2…8 °C
- Description:4′-Hydroxydiclofenac
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results