Antibody package, Protein G
Catalog # 29-0588-06
Supplier: Cytiva
Specifications
- Description:Antibody package, Protein G
- Pk:1 KIT
Specifications
About this item
This Protein G package enables convenient purification of most species of antibodies, including rat.
The HiTrap™ Protein G HP column enclosed is recommended for most species of antibodies, including rat. A HiTrap™ Desalting column is also provided for quick pH neutralisation of the eluted antibodies.
Delivery: Delivery includes: HiTrap™ Protein G HP (1×1 ml) HiTrap™ Desalting (1×5 ml).