Order Entry
Sweden
ContactUsLinkComponent
Sodium hydroxide 25% in aqueous solution, Biotech Reagent, J.T.Baker®
Corrosive
Irritant
Sodium hydroxide 25% in aqueous solution, Biotech Reagent, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Corrosive
Irritant
Sodium hydroxide 25% in aqueous solution, Biotech Reagent, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • GMP Manufactured Product:
    24.5 - 25.5%
  • CAUTION: For Manufacturing, processing or repackaging:
    ≤5 ppm
  • Assay (NaOH):
    24.5 - 25.5 %
  • Chloride (Cl):
    ≤20 ppm
  • Heavy Metals (as Pb):
    ≤1 ppm
  • Iron (Fe):
    ≤2 ppm
  • Endotoxin Concentration (EU/mL):
    ≤2.0

Specification Test Results

Related Information