L-Alanine ≥99%, high purity
Catalog # 0106-VGSK-1KG
Supplier: VWR Collection
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Arsenic:≤0.00015%
- Ash:≤0.15%
- Chloride:≤0.05%
- Heavy Metals:≤0.001%
- Sulphate:0.03%
- Iron:0.003%
- Loss on Drying:0.2%
- pH (5%, Water) @25 °C:5.5 - 7.0
- Purity:99%
- Specific Rotation:13.7 -15.1°
Specification Test Results
About this item
:
:89.09 g/mol
Merck Index:13,00203
Boiling Pt:189 °C (1013 hPa)
Melting Pt:79 °C
Density:1.4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-273-8
CAS Number:56-41-7
Formula:C₃H₇NO₂
MDL Number:MFCD00064410