Order Entry
Sweden
ContactUsLinkComponent
Hexane fraction, ChromAR® for HPLC, Macron Fine Chemicals™
Hexane fraction, ChromAR® for HPLC, Macron Fine Chemicals™
  6751-25
 :  
undefined
Hexane fraction, ChromAR® for HPLC, Macron Fine Chemicals™
  6751-25
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

 

 

  • Assay (GC):
    min. 45 %
  • Residue after Evaporation:
    max. 0.0005 %
  • Titrable Acid (meq/g):
    max. 0.0005
  • Water (KF):
    max. 0.01 %
  • at 210 nm:
    max. 0.2
  • at 220 nm:
    max. 0.1
  • at 230 nm:
    max. 0.02
  • at 240 nm:
    max. 0.01
  • at 250 nm:
    max. 0.004

 

 

 :
 :69 °C (1013 hPa)
 :–94.3 °C
 :0.659 g/cm³ (20 °C)
 :–22 °C
 :Ambient
 :1208
 :3,II
 :GHS02
 :GHS08
 :GHS07
 :GHS09
 :110-54-3