Hexamethylenetetramine ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:100-97-0
:
Formula:C₆H₁₂N₄
MDL Number:MFCD00006895
Merck Index:13,05994
Melting Pt:280 °C
Density:1.33 g/cm³ (20 °C)
Flash Pt:250 °C
Storage Temperature:Ambient
EINECS:202-905-8
ADR:4.1,III
:140.19 g/mol
UN:1328
:GHS02
:GHS07
Specification Test Results
- Assay:Min. 99.0 %
- Heavy metals (as Pb):Max. 10 ppm
- Ignition residue (SO4):Max. 0.05 %
- Water:Max. 1 %
- Cl (Chloride):Max. 20 ppm
- SO4 (Sulphate):Max. 50 ppm