Order Entry
Sweden
ContactUsLinkComponent
Hexamethylenetetramine ≥98%, GPR RECTAPUR®
Hexamethylenetetramine ≥98%, GPR RECTAPUR®
Catalog # 24558.293
Supplier:  VWR Chemicals
CAS Number:  
undefined
Hexamethylenetetramine ≥98%, GPR RECTAPUR®
Catalog # 24558.293
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    Min. 98 %
  • Heavy metals (as Pb):
    Max. 50 ppm
  • Ignition residue (SO4):
    Max. 0.2 %
  • Cl (Chloride):
    Max. 100 ppm
  • SO4 (Sulphate):
    Max. 0.02 %

Specification Test Results

About this item

 :
Merck Index:13,05994
Melting Pt:280 °C
Density:1.33 g/cm³ (20 °C)
Flash Pt:250 °C
Storage Temperature:Ambient
EINECS:202-905-8
UN:1328
ADR:4.1,III
 :140.19 g/mol
 :GHS02
 :GHS07
CAS Number:100-97-0
Formula:C₆H₁₂N₄
MDL Number:MFCD00006895