Order Entry
Sweden
ContactUsLinkComponent
Hexamethylenetetramine, GR for analysis, Supelco®
Hexamethylenetetramine, GR for analysis, Supelco®
Supplier:  Merck
Certificates
undefined
Hexamethylenetetramine, GR for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Assay (perchloric acid titration):
    ≥ 99.5 %
  • Identity (IR-spectrum):
    passes test
  • Appearance:
    colorless or white to almost white crystalline powder
  • pH (100 g/l, water):
    7.0 - 10.0
  • Chloride (Cl):
    ≤ 20 ppm
  • Sulphate (SO₄):
    ≤ 50 ppm
  • Heavy metals (as Pb):
    ≤ 10 ppm
  • NH₄ (Ammonium):
    ≤ 100 ppm
  • Sulfated ash (800 °C):
    ≤ 0.05 %
  • Hazard Class Description:
    Flammable solid

Specification Test Results

Related Information