Order Entry
Sweden
ContactUsLinkComponent
1-Methylpyrrole for synthesis, Sigma-Aldrich®
1-Methylpyrrole for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
1-Methylpyrrole for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    13 g/l (20 °C)
  • Melting Point:
    -57 °C
  • Molar Mass:
    81.12 g/mol
  • Boiling Point:
    111 - 113 °C
  • Flash Point:
    16 °C
  • Refractive Index:
    1.4875 (20 °C, 589 nm)
  • Explosion Limit:
    1.4 - 8.5 %(V)
  • Density:
    0.91 g/cm3 (20 °C)
  • pH:
    10.4 (10 g/l, H2O, 10 °C)
  • Ignition Point:
    400 °C

Specification Test Results

Related Information