Mouse Recombinant NGF-beta (from E. coli)
Supplier: Shenandoah Biotechnology
Certificates
About this item
Nerve growth factor beta (β-NGF) is a neurotrophic factor that is important for the development and maintenance of sensory and sympathetic neurons. β-NGF signals through the low affinity nerve growth factor receptor (LNGFR) and the tropomyosin receptor kinase A (TrkA) to activate PI3K, Ras, and PLC signaling pathways. β-NGF is also involved in the growth, differentiation, and survival of B lymphocytes. Human, mouse, and rat β-NGF proteins are cross-reactive.
- High Quality
- Low Endotoxin
- Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:Bioactivity assay: TF-1 cell proliferation, ED50 ≤5 ng/ml, Bioactivity ≥2,0×10⁵ units/mg
- Gene ID:P01139
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:beta-NGF
- Protein/peptide name:NGF-beta
- Purity:≥95%
- Molecular weight:13,6/27,2 kDa
- Sequence:MSSTHPVFHM GEFSVCDSVS VWVGDKTTAT DIKGKEVTVL AEVNINNSVF RQYFFETKCR ASNPVESGCR GIDSKHWNSY CTTTHTFVKA LTTDEKQAAW RFIRIDTACV CVLSRKATRR G
- Endotoxin level:Low
- Formulation:0,1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay