Order Entry
Sweden
ContactUsLinkComponent
(S)-(-)-Nicotine
(S)-(-)-Nicotine
Catalog # EHERCA15520000
Supplier:  EHRENSTORFER
CAS Number:  
(S)-(-)-Nicotine
Catalog # EHERCA15520000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,5 g
  • Seq:
    3035

Specifications

About this item

 :
Merck Index:12,06611
Boiling Pt:247 °C (1013 hPa)
Melting Pt:–79 °C
Density:1.01 g/cm³ (20 °C)
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:200-193-3
UN:1654
ADR:6.1,II
 :162.23 g/mol
 :GHS06
 :GHS09
CAS Number:54-11-5
Formula:C₁₀H₁₄N₂
MDL Number:MFCD00006369