Mouse Recombinant GM-CSF (from E. coli)
Supplier: Shenandoah Biotechnology
Certificates
About this item
Granulocyte-macrophage colony-stimulating factor (GM-CSF) is hematopoietic growth factor produced by endothelial cells, monocytes, fibroblasts, and T cells. GM-CSF stimulates the production of neutrophilic granulocytes, macrophages, and mixed granulocyte-macrophage colonies from bone marrow cells. GM-CSF promotes immune system development and regulates neutrophil function during infection. Human and mouse GM-CSF show no cross-reactivity.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:Bioactivity Assay: FDCP-1 cell proliferation, ED₅₀ ≤0,05 ng/ml, Bioactivity ≥2,0×10⁷ units/mg
- Gene ID:P01587
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:CSF-2. Pluripoietin-α|mgI-1GM
- Protein/peptide name:GM-CSF
- Purity:≥95%
- Molecular weight:14,3 kDa
- Sequence:MAPTRSPITV TRPWKHVEAI KEALNLLDDM PVTLNEEVEV VSNEFSFKKL TCVQTRLKIF EQGLRGNFTK LKGALNMTAS YYQTYCPPTP ETDCETQVTT YADFIDSLKT FLTDIPFECK KPVQK
- Endotoxin level:Low
- Formulation:10 mM Acetic acid
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay