Human Recombinant SHH (from E. coli)
Sonic hedgehog (SHH) is a member of a small group of hedgehog secreted proteins that are essential for development in both vertebrates and invertebrates. There are three mammalian hedgehog homologues, sonic, desert, and indian, that signal via the Patched-1 and Patched-2 receptors. SHH is a morphogen that is essential during vertebrate organogenesis and adult stem cell division.
- High Quality
- Low Endotoxin
- Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.
: For research use only.
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:Bioactivity assay: CCL-226 cell proliferation, ED50 ≤5 ug/ml, Bioactivity ≥200 units/mg
- Gene ID:Q15465
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:Shh|HHG-1
- Protein/peptide name:SHH
- Purity:≥95%
- Molecular weight:20,2 kDa
- Sequence:MIIGPGRGFG KRRHPKKLTP LAYKQFIPNV AEKTLGASGR YEGKISRNSE RFKELTPNYN PDIIFKDEEN TGADRLMTQR CKDKLNALAI SVMNQWPGVK LRVTEGWDED GHHSEESLHY EGRALDITTS DRDRSKYGML ARLAVEAGFD WVYYESKAHI HCSVKAENSV AAKSGGCFP
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, pH 7,5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay