
6-Chloronicotinic acid ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5326-23-8
:
Formula:C₆H₄ClNO₂
MDL Number:MFCD00006241
:157.56 g/mol
Boiling Pt:284 °C (1013 hPa)
Melting Pt:∼ 192 °C (dec.)
Storage Temperature:Ambient
EINECS:226-201-5
:GHS07