Adenine 99%
Leverantör: Thermo Fisher Scientific
Certifikat
Om denna produkt
CAS-nummer:73-24-5
:
Formel:C₅H₅N₅
MDL-nummer:MFCD00041790
:135.13 g/mol
Merck Index:13,00152
Smält Pt:360…365 °C (dec.)
Densitet:0,99741 g/cm³ (25 °C)
Förvaringstemperatur:Ambient
EINECS:200-796-1
FN:2811
ADR:6.1,III
:GHS06
Specifikationstestresultat
- Appearance:White to yellow Powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 % (105°C, 5 hrs)
- Sulfated ash:≤0.1 %
- HPLC:≥98.5 %
- Titration with HClO4:≥98.5 %