Safety shoes, lace-up, uvex suXXeed®, 62553
Supplier: UVEX
Certificates
About this item
Metal-free, flexible, lightweight safety shoe made from oiled hydro suede leather upper, resistant to liquid penetration, collar and gusset in water-repellent fabric, fabric eyelets plus a double-desity PU2D sole , highly slip resistant and resistant to oils and hydrocarbons.
- Quick drying Climate Airmesh lining, very resistant to abrasion, very good abrasion resistance and very good water vapour absorption
- Removable insole, top in abrasion resistant fabric
- Metal-free protective toecap (200 J)
- Metal-free puncture resistant midsole
Suitable for heavy industry, warehousing and logistics.
Colour: Black/red
Certifications: EN ISO 20345:2011; S3, SRC