Organic reference standard, Phenanthrene 200 µg/ml in isooctane, CIL
Catalog # PROOCILULM74271.2
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:85-01-8
- Description:Phenanthrene 200 µg/ml in isooctane
- Formula:C₁₄H₁₀
- Description:Phenanthrene