Cobalt(II) sulphate heptahydrate ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10026-24-1
:
Formula:CoSO₄·7H₂O
MDL Number:MFCD00149658
:281.1 g/mol
Merck Index:14,2448
Melting Pt:735 °C
Density:2.03
EINECS:233-334-2
UN:3077
ADR:9,III
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient