Order Entry
Sweden
ContactUsLinkComponent
Iron(III) oxide, anhydrous ≥97%, GPR RECTAPUR®
Irritant
Iron(III) oxide, anhydrous ≥97%, GPR RECTAPUR®
Catalog # 24193.292
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Iron(III) oxide, anhydrous ≥97%, GPR RECTAPUR®
Catalog # 24193.292
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    Min. 97 %
  • Cl (Chloride):
    Max. 0.1 %
  • Mg (Magnesium):
    Max. 0.03 %
  • Mn (Manganese):
    Max. 0.3 %
  • Pb (Lead):
    Max. 50 ppm

Specification Test Results

About this item

 :
 :159.69 g/mol
Merck Index:13,04058
Melting Pt:1538 °C
Density:5.24 g/cm³ (20 °C)
EINECS:215-168-2
 :GHS07
CAS Number:1309-37-1
Formula:Fe₂O₃
MDL Number:MFCD00011008